Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032544.3 | NP_115933.2 | 9 | 2395 | ||
GenBank mRNA | AF361482.2 | - | 9 | 2395 | ||
AK010459.1 | - | 2 | 705 | |||
BC055716.1 | - | 9 | 1987 | |||
BC075712.1 | - | 6 | 2423 | |||
BC092042.1 | - | 8 | 1706 | |||
BC138560.1 | - | 9 | 1804 | 8861580 | ||
BC145333.1 | - | 9 | 1801 |