Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Marc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133684.3 | NP_598445.1 | 2 | 452 | ||
GenBank mRNA | AK002874.1 | - | 2 | 450 | ||
AK013440.1 | - | 2 | 472 | |||
AK017150.1 | - | 2 | 452 | |||
AK051230.1 | - | 2 | 453 | |||
AK178160.1 | - | 2 | 278 | |||
AK180823.1 | - | 2 | 278 | |||
BC006888.1 | - | 2 | 435 | 2647757 | ||
BC042686.1 | - | 2 | 456 |