Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039394.1 | NP_001034483.1 | 3 | 992 | ||
NM_133717.3 | NP_598478.1 | 3 | 698 | |||
GenBank mRNA | AK007834.1 | - | 3 | 588 | ||
AK158283.1 | - | 3 | 997 | |||
BC010248.1 | - | 3 | 698 | 3155089 | ||
BY707837.1 | - | 3 | 585 | |||
CV782943.1 | - | 3 | 598 |