Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175124.5 | NP_780333.3 | 8 | 864 | ||
GenBank mRNA | AK006352.1 | - | 2 | 70 | ||
AK077270.1 | - | 8 | 860 | |||
AK159417.1 | - | 8 | 833 | |||
BC027058.1 | - | 5 | 421 | |||
BC052051.1 | - | 6 | 635 | |||
BC086694.1 | - | 8 | 755 | 30697866 |