Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps26b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178027.4 | NP_821170.1 | 6 | 3232 | ||
GenBank mRNA | AK007456.1 | - | 1 | 740 | ||
AK150929.1 | - | 6 | 3153 | |||
AK163860.1 | - | 1 | 935 | |||
AK210283.1 | - | 1 | 34 | |||
AK216567.1 | - | 1 | 285 | |||
AK217669.1 | - | 1 | 285 | |||
AK218652.1 | - | 1 | 285 | |||
BC049180.1 | - | 6 | 3195 | 6314347 | ||
BC052721.1 | - | 6 | 3156 | 6809853 | ||
BC062972.1 | - | 6 | 3156 | 6809878 | ||
BC063783.1 | - | 2 | 1944 |