Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011899.1 | - | 3 | 279 | ||
AK082940.1 | - | 3 | 299 | |||
AK160447.1 | - | 3 | 278 | |||
AK167383.1 | - | 3 | 287 | |||
AK167848.1 | - | 3 | 283 | |||
AY082332.1 | - | 3 | 253 | |||
BC050206.1 | - | 3 | 261 | 30009463 |