Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD51AP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130862.1 | NP_001124334.1 | 10 | 1304 | ||
NM_006479.4 | NP_006470.1 | 9 | 1253 | |||
GenBank mRNA | AF006259.1 | - | 9 | 1148 | ||
AK096930.1 | - | 10 | 1181 | |||
AK290428.1 | - | 10 | 1198 | |||
AK291948.1 | - | 9 | 1147 | |||
AK300773.1 | - | 8 | 1085 | |||
AK309706.1 | - | 3 | 424 | |||
AL832243.1 | - | 10 | 1182 | |||
BC006992.1 | - | 9 | 1147 | 3997352 | ||
BC016330.1 | - | 10 | 1196 | 4107592 |