Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183270.2 | NP_899093.1 | 2 | 296 | ||
GenBank mRNA | AK008883.1 | - | 2 | 262 | ||
AK019895.1 | - | 268 | ||||
AK088786.1 | - | 2 | 284 | |||
AK161506.1 | - | 297 | ||||
BC048574.1 | - | 2 | 237 | |||
BC096605.1 | - | 2 | 263 | 5371531 | ||
BC116995.1 | - | 2 | 265 | 40126314 | ||
BC116997.1 | - | 2 | 265 | 40126316 |