Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wipf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197940.2 | NP_922922.1 | 8 | 2334 | ||
GenBank mRNA | AK046226.1 | - | 1 | 654 | ||
BC057854.1 | - | 8 | 2123 | 6494304 |