Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Piwil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021311.3 | NP_067286.1 | 14, 15 | 1785 | ||
GenBank mRNA | AB032604.1 | - | 14, 15 | 1750 | ||
AF438405.1 | - | 14, 15 | 1785 | |||
BC129857.1 | - | 13, 14 | 1635 | 40109791 | ||
BC129858.1 | - | 13, 14 | 1635 | 40109793 | ||
EF196092.1 | - | 12, 13 | 1595 |