Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF171060.1 | - | 7 | 508 | ||
AK002399.1 | - | 7 | 529 | |||
AK011171.1 | - | 4 | 237 | |||
AK019236.1 | - | 4 | 235 | |||
AK171752.1 | - | 7 | 506 | |||
BC022715.1 | - | 7 | 494 | |||
BC037100.1 | - | 7 | 519 | |||
BC048821.1 | - | 7 | 516 | 3966642 | ||
BG081026.2 | - | 7 | 475 |