Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177795.1 | NP_001171266.1 | 5 | 1016 | ||
NM_001290372.1 | NP_001277301.1 | 4 | 599 | |||
NM_021374.5 | NP_067349.2 | 4 | 669 | |||
GenBank mRNA | AF191552.1 | - | 4 | 559 | ||
AF191554.1 | - | 4 | 545 | |||
AK013773.1 | - | 4 | 498 | |||
AK078208.1 | - | 4 | 514 | |||
AK089947.1 | - | 4 | 489 | |||
AK158864.1 | - | 4 | 510 | |||
BC116925.1 | - | 4 | 559 | 40126244 | ||
BC116929.1 | - | 4 | 559 | 40126248 |