Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prokr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021381.3 | NP_067356.2 | 2 | 343 | ||
GenBank mRNA | AF236082.1 | - | 1 | 97 | ||
AF487278.1 | - | 1 | 97 | |||
AK134279.1 | - | 2 | 357 | |||
AK156734.1 | - | 2 | 517 | |||
BC059003.1 | - | 2 | 343 | 6848525 |
Set Membership: |
G-protein coupled receptor |