Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021480.5 | NP_067455.5 | 3, 4 | 626 | ||
GenBank mRNA | AF134346.2 | - | 3, 4 | 290 | ||
AK077571.1 | - | 3, 4 | 301 | |||
AY116662.1 | - | 2, 3 | 227 | |||
BC058860.1 | - | 3, 4 | 641 | 30014660 |