Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Treh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277847.1 | NP_001264776.1 | 2 | 161 | ||
NM_001277851.1 | NP_001264780.1 | 2 | 165 | |||
NM_021481.3 | NP_067456.1 | 2 | 165 | |||
GenBank mRNA | AF136944.1 | - | 2 | 160 | ||
AK002566.1 | - | 2 | 164 | |||
AK008912.1 | - | 2 | 150 | |||
BC019214.1 | - | 2 | 156 | 5064708 |