Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021500.2 | NP_067475.2 | 3, 4 | 469 | ||
GenBank mRNA | AF263247.1 | - | 3, 4 | 431 | ||
AK007307.1 | - | 3, 4 | 440 | |||
AK034859.1 | - | 3, 4 | 468 | |||
AK049424.1 | - | 3, 4 | 489 | |||
AK075637.1 | - | 2, 3 | 309 | |||
AK088854.1 | - | 3, 4 | 477 | |||
AK145800.1 | - | 3, 4 | 532 | |||
AK152523.1 | - | 3, 4 | 477 | |||
AK168788.1 | - | 3, 4 | 495 | |||
AK169149.1 | - | 3, 4 | 477 | |||
AK183095.1 | - | 1, 2 | 86 | |||
AK185348.1 | - | 1, 2 | 87 | |||
AK202697.1 | - | 1, 2 | 164 | |||
BC039054.1 | - | 3, 4 | 455 | 2648667 | ||
BC058687.1 | - | 3, 4 | 467 | 6411707 |