Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006600.3 | NP_006591.1 | 1, 2 | 213 | ||
XM_017000095.1 | XP_016855584.1 | 1, 2 | 212 | |||
GenBank mRNA | AB019408.1 | - | 1, 2 | 160 | ||
AF086922.1 | - | 1, 2 | 176 | |||
AF100760.1 | - | 1, 2 | 176 | |||
AF125465.1 | - | 1, 2 | 184 | |||
AF130736.1 | - | 1, 2 | 127 | |||
AL136725.1 | - | 1, 2 | 188 | |||
BC002399.2 | - | 1, 2 | 125 | 2822167 | ||
BC003132.1 | - | 1, 2 | 133 | 2958251 | ||
BC006147.1 | - | 1, 2 | 159 | 3954220 | ||
BC007280.1 | - | 1, 2 | 202 | 3343588 | ||
BC015153.1 | - | 1, 2 | 201 | 3883328 | ||
BC021139.1 | - | 1, 2 | 174 | 4871007 | ||
BP348144.1 | - | 1, 2 | 213 |