Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRA1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028045.1 | 2 | 448 | |||
GenBank mRNA | AF047445.1 | - | 2 | 178 | ||
AK290077.1 | - | 1 | 149 | |||
AK304677.1 | - | 2 | 234 | |||
AK310151.1 | - | 2 | 282 | |||
AY334570.1 | - | 1 | 82 | |||
BC069672.1 | - | 2 | 151 | 7262588 | ||
BC069697.1 | - | 2 | 151 | 7262600 | ||
BC069734.1 | - | 2 | 151 | 7262612 | ||
BC101471.1 | - | 2 | 165 | 8068977 | ||
BC101473.1 | - | 2 | 168 | 8068979 | ||
DA776936.1 | - | 2 | 448 |