Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021793.2 | NP_068565.1 | 6 | 1093 | ||
GenBank mRNA | AB045293.1 | - | 6 | 1092 | ||
AK044384.1 | - | 6 | 1094 | |||
AK079893.1 | - | 2 | 240 | |||
BC005722.1 | - | 5 | 969 | 3595205 |