Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271843.1 | NP_001258772.1 | 2 | 353 | ||
NM_022026.3 | NP_071309.1 | 2 | 353 | |||
GenBank mRNA | AB037180.1 | - | 2 | 150 | ||
AK050042.1 | - | 2 | 353 | |||
BC024105.1 | - | 2 | 324 | 5096952 | ||
CT010174.1 | - | 2 | 147 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK050325.1 | - | 1409 | Chr. - - [-] on Build |