Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271843.1 | NP_001258772.1 | 6 | 1114 | ||
NM_022026.3 | NP_071309.1 | 6 | 1114 | |||
GenBank mRNA | AK050042.1 | - | 6 | 1114 | ||
AK154151.1 | - | 6 | 1118 | |||
BC024105.1 | - | 7 | 1235 | 5096952 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK050325.1 | - | 2170 | Chr. - - [-] on Build |