Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl6ip5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022992.2 | NP_075368.1 | 1 | 150 | ||
GenBank mRNA | AF133912.1 | - | 1 | 146 | ||
AF265214.1 | - | 1 | 121 | |||
AF421860.1 | - | 1 | 156 | |||
AK005259.1 | - | 1 | 147 | |||
AK008519.1 | - | 1 | 130 | |||
AK076519.1 | - | 1 | 147 | |||
AK077826.1 | - | 1 | 157 | |||
AK088205.1 | - | 1 | 145 | |||
BC003897.1 | - | 1 | 150 | 3600236 | ||
D87211.1 | - | 1 | 193 |