Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170911.1 | NP_001164382.1 | 4 | 581 | ||
NM_025385.3 | NP_079661.1 | 5 | 699 | |||
GenBank mRNA | AK002430.1 | - | 4 | 549 | ||
AK003850.1 | - | 5 | 696 | |||
AK008611.1 | - | 3 | 521 | |||
AK008894.1 | - | 5 | 659 | |||
AK154583.1 | - | 5 | 659 | |||
AK181565.1 | - | 2 | 356 | |||
AK218697.1 | - | 2 | 249 | |||
AK219155.1 | - | 2 | 319 | |||
BC016234.1 | - | 4 | 845 | 4503179 |