Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lage3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025410.2 | NP_079686.1 | 3 | 593 | ||
GenBank mRNA | AK002539.1 | - | 3 | 594 | ||
AK004210.1 | - | 3 | 586 | |||
AK134624.1 | - | 916 | ||||
BC027305.1 | - | 3 | 559 | 4009230 |