Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lce1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025413.2 | NP_079689.2 | 2 | 708 | ||
GenBank mRNA | AK004290.1 | - | 2 | 709 |