Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nicn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025449.3 | NP_079725.1 | 6 | 1919 | ||
GenBank mRNA | AK075732.1 | - | 6 | 1927 | ||
AK146890.1 | - | 6 | 1929 | |||
AK169135.1 | - | 6 | 1929 | |||
AK195526.1 | - | 1 | 157 | |||
BC024050.1 | - | 6 | 1919 | 5133763 |