Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm4sf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025453.3 | NP_079729.1 | 3 | 384 | ||
GenBank mRNA | AK007532.1 | - | 3 | 385 | ||
AK017154.1 | - | 3 | 386 | |||
AK017195.1 | - | 3 | 385 | |||
AK017209.1 | - | 3 | 393 | |||
BC117736.2 | - | 3 | 376 | 40089849 |