Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCKAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000730.2 | NP_000721.1 | 3 | 641 | ||
GenBank mRNA | AK313978.1 | - | 3 | 683 | ||
AY322549.1 | - | 3 | 488 | |||
BC074987.2 | - | 3 | 536 | 30915519 | ||
L13605.1 | - | 3 | 559 | |||
L19315.1 | - | 3 | 641 |
Set Membership: |
G-protein coupled receptor |