Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rmdn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025476.6 | NP_079752.3 | 10 | 1651 | ||
GenBank mRNA | AK029446.1 | - | 10 | 1510 |