Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025588.2 | NP_079864.1 | 2, 3 | 214 | ||
GenBank mRNA | AK016532.1 | - | 3, 4 | 341 | ||
AK032766.1 | - | 2, 3 | 236 | |||
AK042744.1 | - | 2, 3 | 261 | |||
AK050068.1 | - | 2, 3 | 222 | |||
AK180355.1 | - | 1, 2 | 107 | |||
BC048154.1 | - | 2, 3 | 214 | 6400932 | ||
BC049102.1 | - | 2, 3 | 214 | 5702581 |