Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145158.1 | NP_001138630.1 | 3 | 359 | ||
NM_001145207.1 | NP_001138679.1 | 4 | 485 | |||
NM_006620.3 | NP_006611.1 | 4 | 485 | |||
XM_017010192.1 | XP_016865681.1 | 3 | 287 | |||
GenBank mRNA | AF161394.1 | - | 4 | 410 | ||
AJ420551.1 | - | 4 | 402 | |||
AJ459826.1 | - | 4 | 471 | |||
AJ459827.1 | - | 4 | 471 | |||
AK024258.1 | - | 4 | 406 | |||
AK295545.1 | - | 3 | 280 | |||
BC001465.2 | - | 4 | 394 | 3138910 | ||
BC040849.1 | - | 4 | 389 | 5104748 | ||
DB051030.1 | - | 4 | 486 | |||
U87791.1 | - | 4 | 472 |