Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamtor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025605.3 | NP_079881.2 | 2, 3 | 354 | ||
GenBank mRNA | AK003509.1 | - | 2, 3 | 246 | ||
AK009320.1 | - | 2, 3 | 243 | |||
AK010247.1 | - | 2, 3 | 287 | |||
AK013389.1 | - | 2, 3 | 264 | |||
AK019947.1 | - | 2, 3 | 205 | |||
AK144476.1 | - | 2, 3 | 253 | |||
AK203171.1 | - | 2, 3 | 196 | |||
BC020142.1 | - | 2, 3 | 260 | 3709051 | ||
BC092062.1 | - | 2, 3 | 246 | 4024506 | ||
BC096412.1 | - | 2, 3 | 226 | 5255882 | ||
BC126973.1 | - | 2, 3 | 261 | 40100355 |