Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025616.3 | NP_079892.1 | 2 | 264 | ||
GenBank mRNA | AK008340.1 | - | 2 | 265 | ||
AK008382.1 | - | 2 | 262 | |||
AK010832.1 | - | 2 | 230 | |||
AK012977.1 | - | 2 | 261 | |||
AK088074.1 | - | 2 | 257 | |||
AK167473.1 | - | 2 | 267 | |||
AK178698.1 | - | 1 | 38 | |||
AY551342.1 | - | 2 | 250 | |||
BC010303.1 | - | 2 | 239 | 3597676 | ||
BC037056.1 | - | 2 | 266 |