Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sike1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025679.3 | NP_079955.1 | 5 | 1335 | ||
GenBank mRNA | AK017694.1 | - | 5 | 1331 | ||
AK033553.1 | - | 5 | 1339 | |||
AK035590.1 | - | 5 | 1349 |