Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037726.1 | NP_001032815.1 | 7 | 4964 | ||
NM_009952.2 | NP_034082.1 | 9 | 5128 | |||
NM_133828.2 | NP_598589.2 | 8 | 5086 | |||
GenBank mRNA | AK014391.1 | - | 1 | 1597 | ||
AK032372.1 | - | 1 | 1579 | |||
BC048151.1 | - | 1 | 1352 | |||
BC058366.1 | - | 1 | 1352 | |||
BC071201.1 | - | 1 | 1784 |