Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025730.3 | NP_080006.3 | 41 | 6027 | ||
GenBank mRNA | AK014886.1 | - | 2 | 86 | ||
AK014938.1 | - | 2 | 99 | |||
AK034413.1 | - | 18 | 2650 | |||
AY792512.1 | - | 41 | 5970 | |||
BC034074.1 | - | 7 | 910 |