Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025748.4 | NP_080024.3 | 5, 6 | 892 | ||
GenBank mRNA | AK016938.1 | - | 5, 6 | 907 | ||
AK021291.1 | - | 5, 6 | 580 | |||
BC062195.1 | - | 5, 6 | 610 | 5328466 |