Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006672.3 | NP_006663.2 | 9 | 1676 | ||
NM_153320.2 | NP_696961.2 | 10 | 1682 | |||
XM_006714970.3 | XP_006715033.1 | 10 | 1691 | |||
XM_006714971.3 | XP_006715034.1 | 9 | 1685 | |||
XM_011514256.2 | XP_011512558.1 | 10 | 1868 | |||
XM_011514257.2 | XP_011512559.1 | 10 | 1859 | |||
XM_011514261.2 | XP_011512563.1 | 11 | 1633 | |||
XM_011514263.2 | XP_011512565.1 | 10 | 1435 | |||
XM_017010199.1 | XP_016865688.1 | 9 | 1709 | |||
XM_017010200.1 | XP_016865689.1 | 11 | 1673 | |||
XM_017010201.1 | XP_016865690.1 | 11 | 1643 | |||
GenBank mRNA | AF097518.1 | - | 10 | 1678 | ||
AK290796.1 | - | 9 | 1595 | |||
AY050498.1 | - | 10 | 1583 | |||
BC033805.1 | - | 10 | 1603 | 5187472 |