Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040662.1 | 2 | 659 | |||
GenBank mRNA | AK093953.1 | - | 4 | 544 | ||
AK290875.1 | - | 2 | 654 | |||
BC106759.2 | - | 2 | 605 | 40024782 | ||
BC113853.1 | - | 1 | 696 | |||
BC114462.1 | - | 2 | 605 | |||
BX640979.1 | - | 2 | 657 | |||
L06175.1 | - | 2 | 651 |