Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lonp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025827.3 | NP_080103.1 | 4 | 848 | ||
GenBank mRNA | AK004843.1 | - | 4 | 762 | ||
AK050231.1 | - | 4 | 757 | |||
AK088348.1 | - | 4 | 849 | |||
AK151103.1 | - | 4 | 794 | |||
AK168791.1 | - | 4 | 794 | |||
AK168956.1 | - | 4 | 794 | |||
AK169656.1 | - | 762 | ||||
AK218948.1 | - | 1 | 113 | |||
BC049090.1 | - | 4 | 784 | 6401542 |