Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt16l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025839.3 | NP_080115.1 | 3 | 839 | ||
GenBank mRNA | AF435792.1 | - | 3 | 851 | ||
AK002378.1 | - | 3 | 832 | |||
AK003241.1 | - | 3 | 840 | |||
AK007897.1 | - | 3 | 835 | |||
AK019936.1 | - | 4 | 727 | |||
AK159841.1 | - | 3 | 832 | |||
AK183937.1 | - | 1 | 187 | |||
AK193716.1 | - | 1 | 187 | |||
BC025569.1 | - | 3 | 913 | 5322456 |