Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025853.3 | NP_080129.2 | 11 | 1841 | ||
GenBank mRNA | AK006246.1 | - | 11 | 1842 | ||
AK017740.1 | - | 11 | 1842 | |||
AK017813.1 | - | 11 | 1796 | |||
AK145373.1 | - | 12 | 1669 | |||
BC025079.1 | - | 11 | 1824 | 3499844 | ||
BC046807.1 | - | 11 | 1802 | 6829342 | ||
CK333431.1 | - | 1 | 85 |