Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027349.3 | NP_081625.3 | 19 | 3770 | ||
GenBank mRNA | AK076553.1 | - | 5 | 1898 | ||
AK087936.1 | - | 7 | 2096 | |||
AK142132.1 | - | 13 | 3046 | |||
BC066150.1 | - | 8 | 2203 | |||
BC067400.1 | - | 8 | 2203 | |||
BC158104.1 | - | 18 | 3650 | 9007388 |