Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sectm1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026907.3 | NP_081183.3 | 4, 5 | 629 | ||
GenBank mRNA | AK007321.1 | - | 3, 4 | 491 | ||
AK029082.1 | - | 4, 5 | 630 | |||
AK143807.1 | - | 4, 5 | 613 | |||
BC020159.1 | - | 3, 4 | 580 | 4016267 |