Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ghitm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199122.1 | NP_001186051.1 | 2 | 330 | ||
NM_078478.5 | NP_510963.1 | 2 | 283 | |||
GenBank mRNA | AF412297.1 | - | 2 | 227 | ||
AK005636.1 | - | 2 | 227 | |||
AK093887.1 | - | 2 | 259 | |||
AK134592.1 | - | 2 | 213 | |||
AK146433.1 | - | 2 | 190 | |||
AK146853.1 | - | 2 | 246 | |||
AK151488.1 | - | 2 | 260 | |||
AK158904.1 | - | 2 | 273 | |||
AK159695.1 | - | 2 | 257 | |||
AK159724.1 | - | 2 | 257 | |||
AK164122.1 | - | 2 | 228 | |||
AK167970.1 | - | 2 | 254 | |||
AK168022.1 | - | 2 | 260 | |||
AK168678.1 | - | 2 | 331 | |||
AK168800.1 | - | 2 | 239 | |||
BC008622.1 | - | 2 | 261 | 3588758 | ||
BC010224.1 | - | 2 | 263 | 3500685 | ||
BY141813.1 | - | 2 | 289 |