Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080554.2 | NP_542121.2 | 10 | 1399 | ||
GenBank mRNA | AK005335.1 | - | 10 | 1381 | ||
AK011026.1 | - | 10 | 1376 | |||
AK029067.1 | - | 8 | 2776 | |||
AK032992.1 | - | 10 | 1377 | |||
AK078419.1 | - | 10 | 1377 | |||
AK145587.1 | - | 10 | 1376 | |||
AK148284.1 | - | 1400 | ||||
AK208538.1 | - | 2 | 223 | |||
AK220321.1 | - | 10 | 1340 | |||
BC010650.1 | - | 4 | 500 | |||
BC019112.1 | - | 10 | 1410 |