Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319676.1 | NP_001306605.1 | 12 | 1552 | ||
NM_006615.2 | NP_006606.1 | 14 | 1741 | |||
NM_016452.2 | NP_057536.1 | 13 | 1663 | |||
XM_011544017.1 | XP_011542319.1 | 14 | 1725 | |||
XM_011544018.1 | XP_011542320.1 | 13 | 1647 | |||
XM_011544019.2 | XP_011542321.1 | 12 | 1647 | |||
XM_011544020.1 | XP_011542322.1 | 14 | 1471 | |||
XM_017000098.1 | XP_016855587.1 | 13 | 1641 | |||
XM_017000099.1 | XP_016855588.1 | 14 | 1471 | |||
GenBank mRNA | AB038463.1 | - | 13 | 1632 | ||
AF022799.1 | - | 14 | 1741 | |||
AK298024.1 | - | 12 | 1491 | |||
AK309039.1 | - | 15 | 1655 | |||
AK313629.1 | - | 14 | 1725 | |||
BC027993.1 | - | 12 | 1503 | 5173512 |