Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178922.3 | NP_849253.2 | 3 | 1704 | ||
GenBank mRNA | AF117382.1 | - | 1 | 428 | ||
AJ868292.1 | - | 2 | 1668 | |||
AK028357.1 | - | 2 | 1516 | |||
AK084975.1 | - | 3 | 1627 | |||
AK129266.1 | - | 3 | 1690 | |||
AK145386.1 | - | 1 | 811 | |||
AK156332.1 | - | 3 | 1720 | |||
AK157276.1 | - | 3 | 1720 | |||
BC065124.1 | - | 1703 | 30537019 |