Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2610001J05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002763.1 | - | 3 | 242 | ||
AK011259.1 | - | 3 | 243 | |||
AK028169.1 | - | 3 | 261 | |||
AK035027.1 | - | 3 | 243 | |||
AK136158.1 | - | 3 | 244 | |||
AK152937.1 | - | 296 | ||||
AW611020.1 | - | 3 | 261 | |||
BC070405.1 | - | 2 | 227 |