Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naglu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013792.2 | NP_038820.2 | 3 | 587 | ||
GenBank mRNA | AK143546.1 | - | 3 | 588 | ||
AK158045.1 | - | 1 | 1805 | |||
AK158774.1 | - | 2 | 1383 | |||
AK164125.1 | - | 3 | 566 | |||
AK170758.1 | - | 3 | 570 | |||
BC055733.1 | - | 3 | 561 | 6411530 | ||
U85247.1 | - | 3 | 556 |